Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC5414275474

Accela ChemBio Inc.

6-Benzyl-1,2,3,4-tetrahydro-6H-pyrrolo[3,4-b]pyridine-5,7-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
242.28
MDL number:

Iupac Name

6-benzyl-3,4-dihydro-1H-pyrrolo[3,4-b]pyridine-5,7(2H,6H)-dione

Inchi Code

1S/C14H14N2O2/c17-13-11-7-4-8-15-12(11)14(18)16(13)9-10-5-2-1-3-6-10/h1-3,5-6,15H,4,7-9H2

InChI key

JJFNYKMZELJLBL-UHFFFAOYSA-N

storage temp.

+4C

Purity

95%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

White to orange solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY005506&attname=GoodsCode

MSDS

http://www.accelachem.com/cn/msds/SY005506.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service