AC5388701967
Accela ChemBio Inc.
2-Chloro-4-(diethylamino)pyrido[3,2-d]pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-chloro-N,N-diethylpyrido[3,2-d]pyrimidin-4-amine
Inchi Code
1S/C11H13ClN4/c1-3-16(4-2)10-9-8(6-5-7-13-9)14-11(12)15-10/h5-7H,3-4H2,1-2H3
InChI key
BVRSXOHTYPPHOK-UHFFFAOYSA-N
storage temp.
+4C
Purity
97%
Country of Origin
CN
Shipping Temperature
RT
Physical Form
White to brown solid
Product Link
http://www.accelachem.com/cn/search.html?keyword=SY022728&attname=GoodsCode
MSDS
http://www.accelachem.com/cn/msds/SY022728.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service