Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC5372909511

Accela ChemBio Inc.

3,6-Dimethyl-2-nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
167.16
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3,6-dimethyl-2-nitrophenol

Inchi Code

1S/C8H9NO3/c1-5-3-4-6(2)8(10)7(5)9(11)12/h3-4,10H,1-2H3

InChI key

VIQHHRZADKSPIM-UHFFFAOYSA-N

storage temp.

+4C

Purity

97%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

Light yellow to orange solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY005501&attname=GoodsCode

pictograms

MSDS

http://accelachem.com/cn/msds/SY005501.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service