Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH99A47437

AA BLOCKS, INC.

2-bromo-4-ethyl-1,3-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
192.08
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

2-bromo-4-ethyl-1,3-thiazole

Inchi Code

1S/C5H6BrNS/c1-2-4-3-8-5(6)7-4/h3H,2H2,1H3

InChI key

YQWKKYUKBQHMJG-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/89322-56-5

Synonym

2-bromo-4-ethyl-1,3-thiazole

MSDS

https://www.aablocks.com/downloadSds?cas=89322-56-5


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service