Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH99A43EEF

AA BLOCKS, INC.

2-Methoxy-3-(3-methyl-2-thenoyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.29
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

(2-methoxy-3-pyridinyl)(3-methyl-2-thienyl)methanone

Inchi Code

1S/C12H11NO2S/c1-8-5-7-16-11(8)10(14)9-4-3-6-13-12(9)15-2/h3-7H,1-2H3

InChI key

HXUHMWJDLFEGPW-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

97%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/898785-87-0

Synonym

2-Methoxy-3-(3-methyl-2-thenoyl)pyridine97%

MSDS

https://www.aablocks.com/downloadSds?cas=898785-87-0


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service