Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH97CE8E92

AA BLOCKS, INC.

2-mercapto-N-[3-(trifluoromethyl)phenyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
235.23
MDL number:

Iupac Name

2-sulfanyl-N-[3-(trifluoromethyl)phenyl]acetamide

Inchi Code

1S/C9H8F3NOS/c10-9(11,12)6-2-1-3-7(4-6)13-8(14)5-15/h1-4,15H,5H2,(H,13,14)

InChI key

LRIKARNWWZHGBR-UHFFFAOYSA-N

storage temp.

2~8 C

Purity

95%

Country of Origin

US

Product Link

https://www.aablocks.com/prod/34282-26-3

Synonym

2-sulfanyl-N-[3-(trifluoromethyl)phenyl]acetamide

MSDS

https://www.aablocks.com/downloadSds?cas=34282-26-3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service