Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH97CD7F9F

AA BLOCKS, INC.

1H-1,3-benzodiazole; perylene

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$107.90
100 MG
$344.50

Vendor SKU: AA008LUJ-25MG

$107.90


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$107.90
100 MG
$344.50

About This Item

CAS Number:
Molecular Weight:
370.45
MDL number:

Vendor SKU: AA008LUJ-25MG

$107.90


Ships in 21 days - from Aldrich Partner.

Iupac Name

1H-benzo[d]imidazole--perylene (1/1)

Inchi Code

1S/C20H12.C7H6N2/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17;1-2-4-7-6(3-1)8-5-9-7/h1-12H;1-5H,(H,8,9)

InChI key

KIIFVSJBFGYDFV-UHFFFAOYSA-N

storage temp.

2~8 C

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/79534-91-1

Synonym

PTCBI (cis- and trans- mixture)

MSDS

https://www.aablocks.com/downloadSds?cas=79534-91-1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service