Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AABH303718D6

AA BLOCKS, INC.

(R)-2,5,8-Trimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$523.90

Vendor SKU: AA001X01-100MG

$523.90


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$523.90

About This Item

CAS Number:
Molecular Weight:
416.69
MDL number:

Vendor SKU: AA001X01-100MG

$523.90


Ships in 10 days - from Aldrich Partner.

Iupac Name

(R)-2,5,8-trimethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol

Inchi Code

1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1

Inchi Key

WGVKWNUPNGFDFJ-DQCZWYHMSA-N

Storage temp.

2~8C

Purity

99%

Country of Origin

CN

Product Link

https://www.aablocks.com/prod/16698-35-4

Synonym

[2R[2R*(4R*,8R*)]]-3,4-dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol

MSDS

https://www.aablocks.com/downloadSds?cas=16698-35-4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service