Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS2045

Clethodim

analytical standard

Synonym(s):

2-{(E)-1-[(E)-3-Chloroallyloxyimino]propyl}-5-[2-(ethylthio)propyl]-3-hydroxy-2-cyclohexen-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26ClNO3S
CAS Number:
Molecular Weight:
359.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-2045

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCSC(C)CC1CC(O)=C(C(=O)C1)\C(CC)=N\OC\C=C\Cl

InChI

1S/C17H26ClNO3S/c1-4-14(19-22-8-6-7-18)17-15(20)10-13(11-16(17)21)9-12(3)23-5-2/h6-7,12-13,20H,4-5,8-11H2,1-3H3/b7-6+,19-14+

InChI key

SILSDTWXNBZOGF-KUZBFYBWSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Clethodim is a cyclohexanedione oxime herbicide primarily used against a wide variety of broadleaf crops. Its mode of action involves the inhibition of acetyl-coenzyme A carboxylase.[1]

Application

Clethodim may be used as a reference standard for the determination of clethodim in crops by liquid chromatography (LC) followed by liquid chromatography/mass spectrometry (LC/MS) with electrospray ionization (ESI).[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Skin Sens. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physiological basis for antagonism of clethodim by imazapic on goosegrass (Eleusine indica (L.) Gaertn.).
Burke LC and Wilcut JW
Pesticide Biochemistry and Physiology, 76(2), 37-45 (2003)
Theola Louie et al.
Bioorganic & medicinal chemistry letters, 20(15), 4611-4613 (2010-06-29)
A series of dimeric 1,3-cyclohexanedione oxime ethers were synthesized and found to have significant antiplasmodial activity with IC(50)'s in the range 3-12 microM. The most active dimer was tested in the Plasmodium berghei mouse model of malaria and at a
S Ishimitsu et al.
Journal of AOAC International, 84(4), 1172-1178 (2001-08-15)
A method was developed for determination of the herbicide clethodim (C0) and its oxidation metabolites clethodim sulfoxide (C1) and clethodim sulfone (C2) in agricultural products. Upon extraction, both C0 and C1 were oxidized to C2 by m-chloroperoxybenzoic acid, and C2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service