Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AR2026

Supelco

Oatmeal (C, H, N, S)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

manufacturer/tradename

Alpha Resources

application(s)

food and beverages

format

matrix material

General description

Oatmeal (C, H, N, S) is a reference standard used for the verification of food or feed analysis by various test methods. This reference has an indefinite known shelf life unopened. Once opened this certificate is limited to two years. Keep sealed and store under normal laboratory conditions. This reference standard was produced in accordance to ISO Guide 31 and ISO 17034.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Carbon, Hydrogen, Nitrogen and Sulphur

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service