Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S8540

Sigma-Aldrich

Ser-Gly-Gln-Ser-Trp-Arg-Pro-Gln-Gly-Arg-Phe-NH2

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H85N21O15
CAS Number:
Molecular Weight:
1304.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

NC(CO)C(=O)NCC(=O)NC(CCC(N)=O)C(=O)NC(CO)C(=O)NC(Cc1c[nH]c2ccccc12)C(=O)NC(CCCNC(N)=N)C(=O)N3CCCC3C(=O)NC(CCC(N)=O)C(=O)NCC(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4ccccc4)C(N)=O

Gene Information

human ... NPY(4852)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cardioexcitatory peptide from the African giant snail

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pedder et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 103(3), 441-451 (1992-11-01)
1. Intracellular recordings were made from identified neurones in the suboesophageal ganglia of Helix aspersa. Seven neuropeptides were tested for activity and their actions compared with acetylcholine and FMRFamide. 2. Three peptides isolated from nematodes, AF-1, AF-2 and PAN-1 had
M Fujimoto et al.
Biochemical and biophysical research communications, 242(2), 436-440 (1998-02-03)
A novel bioactive peptide with the C-terminal RFamide was isolated from the brain of the Japanese crucian carp, Carassius auratus langsdorfii, by using the intestine of the fish as the bioassay system. The primary structure of the peptide was determined
K Fujimoto et al.
Biochemical and biophysical research communications, 167(2), 777-783 (1990-03-16)
An undecapeptide which potentiates the beat of the ventricle in the African giant snail, Achatina fulica Ferussac, was purified from the atria of the snail. Its primary structure was determined to be H-Ser-Gly-Gln-Ser-Trp-Arg-Pro-Gln-Gly-Arg-Phe-NH2. This peptide was found to have excitatory
C Baud et al.
Cellular and molecular neurobiology, 18(4), 413-424 (1998-06-10)
1. We report the isolation of a peptide from the brain of the snail Helix aspersa by radioimmunoassay using an antisomatostatin. 2. The sequencing of an immunopositive fraction showed the presence of a new tridecapeptide, termed Helix cardioinhibitory peptide (HCIP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service