S2140
Sodium formate
ACS reagent, ≥99.0%
Synonym(s):
Formic acid sodium salt
About This Item
Recommended Products
grade
ACS reagent
assay
≥99.0%
impurities
≤0.005% Insoluble matter
mp
259-262 °C (lit.)
anion traces
chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.001%
cation traces
Ca: ≤0.005%
Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm
SMILES string
[Na+].[O-]C=O
InChI
1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key
HLBBKKJFGFRGMU-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service