Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N1160

Sigma-Aldrich

Norbiotinamine hydrochloride

≥98% (Vendor information, TLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18ClN3OS
Molecular Weight:
251.78
MDL number:
UNSPSC Code:
12352204

assay

≥98% (Vendor information, TLC)

form

solid

storage temp.

room temp

Application

Used to measure tissue transglutaminase activity.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Smethurst et al.
The Biochemical journal, 313 ( Pt 3), 803-808 (1996-02-01)
Electropermeabilized human endothelial cells (ECV-304) were used to study the regulation of tissue transglutaminase (tTGase) activity in the intracellular environment. An ELSA (enzyme-linked sorbent assay) plate assay was developed for intracellular tTGase activity, using the incorporation of a biotinylated primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service