MAK027
Nickel Assay Kit
sufficient for 100 colorimetric tests
About This Item
Recommended Products
usage
sufficient for 100 colorimetric tests
detection method
colorimetric
storage temp.
2-8°C
General description
In the Nickel Assay kit, Ni2+ reacts with 2-mercaptoethanol in borate buffer to form a complex with strong absorbance bands from 300 to 600 nm. Fe2+ and Co2+ interfere with the assay; therefore, extra steps must be taken to subtract the interference in order to determine the correct nickel concentration in mixed samples. Other ions tested (Mn2+, Cu2+, and Zn2+) do not interfere with the assay and presumably no other ionic species will be present in high enough concentration to interfere with the reaction. The assay is a simple method of quantifying Ni2+ in a variety of samples, which gives a linear range of 2–50 nmole nickel in samples containing <25 nmole of Co2+ ions.
Suitability
Principle
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Oral
target_organs
Liver,Heart, Lungs
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service