Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK005

Sigma-Aldrich

Phenylalanine Assay Kit

sufficient for 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

neonatal diseases; neurological disorders; pediatric diseases

storage temp.

−20°C

General description

Phenylalanine is a non-polar essential amino acid. In the liver, phenylalanine is converted to tyrosine, which is a precursor for multiple compounds including L-DOPA and melanin. Defects in the activity of phenylalanine hydroxylase result in the inherited metabolic disorder phenylketonuria (PKU). PKU results in the build up of phenylalanine and phenylalanine metabolites, and can result in growth defects and mental retardation if not treated.

Suitability

Suitable for L-phenylalanine detection in cell and tissue culture supernatants, serum, and other biological samples

Principle

Phenylalanine concentration is determined by a coupled enzyme assay, which results in the deamination of phenylalanine and the production of NADH which reacts with the probe resulting in a fluorescent (λex = 535 nm/λem = 587 nm) product, proportional to the phenylalanine present.

replaced by

Product No.
Description
Pricing

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Crook et al.
ACS synthetic biology, 9(5), 1010-1021 (2020-04-24)
The development of robust engineered probiotic therapies demands accurate knowledge of genetic construct expression in the gut. However, the monetary and ethical costs of testing engineered strains in vertebrate hosts are incompatible with current high-throughput design-build-test cycles. To enable parallel
AMINO ACIDS | Metabolism
Encyclopedia of Food Sciences and Nutrition (Second Edition) (2003)
Phenylketonuria: an inborn error of phenylalanine metabolism.
Williams R A, et al.
The Clinical Biochemist. Reviews / Australian Association of Clinical Biochemists, 29(1), 31-31 (2008)
Protein and Amino Acid Metabolism
Essentials of Medical Biochemistry null
CTI Reviews
Clinical Chemistry, Principles, Procedures, Correlations (2016)

Protocols

Assay protocol for the fluorometric detection of Phenylalanine in biological samples using the phenylalanine assay kit.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service