Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L7213

Sigma-Aldrich

Collagen G

Type 1 from calf skin, 0.4% solution in HCl, 4 mg/mL

Synonym(s):

Collagen Gel, Collagen Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine (calf) skin

form

liquid

packaging

100 mL of

concentration

4 mg/mL

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

storage temp.

2-8°C

General description

This product is identical to Biochrom GmbH part number L 7213. It is comprised of acid-soluble calfskin collagens in a 0.4 % solution containing 15 mmol/l HCl. Collagen G is used in the production of gels, to embed cells. Collagen G gels are suitable for use as a substrate for adherent cells in a cell culture vessel and as a floating matrix in or on cell culture medium.

Preparation Note

Recommendations for optimum gelling with Collagen G
Geling is heavily influenced by the pH value. Before starting work, pre-cool all reagents to a temperature of +2 - +8C.

Solution A:
0.7 M sodium hydroxide and 1 M HEPES buffer are mixed equally (e.g. 5 ml sodium hydroxide with 5 ml HEPES buffer).

Solution B:
A 10x medium (e.g. RPMI 1640) and solution A are mixed equally (e.g. 5 ml medium with 5 ml solution A).

  • The pH of solution B should be between 7.90 and 8.05. It is advisable to check the pH using a suitable measuring method. If the gel should remain sterile, measure the pH of an aliquot previously removed.
  • 8.0 ml Collagen G is gently mixed with 2.0 ml of solution B to produce the ready-to-use solution for gelling. This prevents the formation of air bubbles.
  • 3.0 ml of solution prepared in this way is pipetted into a 25 cm2 culture flask. For a culture flask with a diameter of 9 cm, 5.0 ml is required. The culture flask is then incubated vibration-free for at least 1 hour or, for optimum stability, 24 hours at +35°C (± 2°C).

Examining the gel produced under a microscope and taking pictures
If a 35 mm culture dish is filled with 2 ml Collagen G/buffer media mixture (pH approx. 7.2 - 7.8) and incubated, a gel with faint white diffusion is formed, viewed from above. This diffusion appears as a result of crosslinking of the collagen molecules and should not cause any problems when examining under a microscope or taking pictures.

The accumulation of diffusion is proportional to the increase of thickness of the gel, for example if 5-7 ml instead of 2 ml per 10 cm2 Collagen G (35 mm culture vessel) is used. This could make microscopic and photographic analysis difficult.

Other Notes

Specifications:

Cell Attachment: Pass
pH: 1.8-2.4
Osmolality: 0.01-0.05 Osm/kg H20
Collagen Content: 0.37-0.45%
Hydroxyproline: 0.046-0.0565%
Color: Colorless
Appearance: Clear

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service