Skip to Content
MilliporeSigma
All Photos(1)

Documents

G3266

Sigma-Aldrich

α-D-Galactopyranosylphenyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO6S
CAS Number:
Molecular Weight:
313.33
MDL number:
UNSPSC Code:
12352201

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](Oc2ccc(cc2)N=C=S)[C@H](O)[C@@H](O)[C@H]1O

Application

Suitable for the preparation of neoglycoproteins.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Monsigny et al.
Biology of the cell, 51(2), 187-196 (1984-01-01)
The presence and the sugar specificity of membrane lectins on the cell surface of mouse L1210 leukemia cells were demonstrated by using various neoglycoproteins (glycosylated serum albumin) substituted with fluorescein or methotrexate. Neoglycoproteins were prepared by reaction of glycosidophenylisothiocyanates with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service