C2902
2-Cyano-6-methoxybenzothiazole
Synonym(s):
6-Methoxy-2-benzothiazolecarbonitrile
About This Item
mp
129-131 °C (lit.)
SMILES string
COc1ccc2nc(sc2c1)C#N
InChI
1S/C9H6N2OS/c1-12-6-2-3-7-8(4-6)13-9(5-10)11-7/h2-4H,1H3
InChI key
DEWDWBYQOFXKIH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
comparable product
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service