Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51581

Sigma-Aldrich

2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium salt

≥75% (TLC)

Synonym(s):

(2S,8R,18Z)- 2-Amino-5-hydroxy-10-oxo-8-[[(1-oxohexadecyl)oxy]methyl]-4,6,9-trioxa-5-phosphaheptacos-18-enoic acid 5-oxide monosodium salt, 1-Hexadecanoyl-2-(cis-9-octadecenoyl)-sn-glycero-3-phospho-L-serine sodium salt, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine sodium salt, 3-sn-Phosphatidyl-L-serine, 2-oleoyl-1-palmitoyl sodium salt, POPS-Na, PS(16:0/18:1(9Z)), PS(16:0/18:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H75NNaO10P
CAS Number:
Molecular Weight:
783.99
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

assay

≥75% (TLC)

form

powder or crystals

functional group

amine
ester

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OC[C@H](N)C(O)=O)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C40H76NO10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(43)51-36(34-49-52(46,47)50-35-37(41)40(44)45)33-48-38(42)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,36-37H,3-16,19-35,41H2,1-2H3,(H,44,45)(H,46,47);/q;+1/p-1/b18-17-;/t36-,37+;/m1./s1

InChI key

RQYKXRYGZHMUSU-JEAFVVATSA-M

Application

Forms negatively charged lipid bilayers. A molecular dynamics simulation indicates that Na+ ions penetrate unexpectedly far into the bilayer, well beyond the hydrophilic end groups.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Marc A Sprague-Piercy et al.
Biomolecules, 10(7) (2020-07-28)
The Droserasins, aspartic proteases from the carnivorous plant Drosera capensis, contain a 100-residue plant-specific insert (PSI) that is post-translationally cleaved and independently acts as an antimicrobial peptide. PSIs are of interest not only for their inhibition of microbial growth, but
Michele F M Sciacca et al.
Biochimica et biophysica acta. Biomembranes (2018-03-21)
Type II diabetes mellitus (T2DM) is characterized by the presence of amyloid deposits of the human islet amyloid polypeptide (hIAPP) in pancreatic β-cells. A wealth of data supports the hypothesis that hIAPP's toxicity is related to an abnormal interaction of
Carina Drechsler et al.
Biophysical journal, 115(8), 1509-1517 (2018-09-30)
Lipid asymmetries between the outer and inner leaflet of the lipid bilayer exist in nearly all biological membranes. Although living cells spend great effort to adjust and maintain these asymmetries, little is known about the biophysical phenomena within asymmetric membranes
Yannick R F Schmid et al.
Advanced biosystems, 4(11), e2000153-e2000153 (2020-10-22)
Giant unilamellar lipid vesicles (GUVs) are widely used as model membrane systems and provide an excellent basis to construct artificial cells. To construct more sophisticated artificial cells, proteins-in particular membrane proteins-need to be incorporated in GUVs. However, current methods for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service