24105
Ethanol (denatured)
96.0-97.2%, puriss., meets analytical specification of Ph. Eur., BP
Synonym(s):
Ethyl alcohol
About This Item
Recommended Products
Product Name
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
vapor density
1.59 (vs air)
Quality Level
vapor pressure
44.6 mmHg ( 20 °C)
grade
puriss.
assay
96.0-97.2%
form
liquid
autoignition temp.
683 °F
quality
meets analytical specification of Ph. Eur., BP
expl. lim.
19 %, 60 °F
impurities
Residual solvents, complies
acid. or alk. react. impurities, in accordance
volatile impurities, in accordance
≤0.001% non-volatile matter
≤0.02% methanol (GC)
4.8-7.4% water (Karl Fischer)
refractive index
n20/D 1.3600 (lit.)
pH
7.0 (20 °C, 10 g/L)
bp
78 °C (lit.)
mp
−114 °C (lit.)
density
0.805-0.812 g/mL at 20 °C
absorption
in accordance
suitability
complies for appearance
complies for identity A
complies for identity B
format
neat
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Caution
Footnote
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
55.4 °F - closed cup
flash_point_c
13 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service