Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54580

Sigma-Aldrich

3-Hydroxybenzaldehyde

purum, ≥95.0% (HPLC), faintly yellow to light brown

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein/REAXYS Number:
507099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥95.0% (HPLC)

form

solid

color

faintly yellow to light brown

bp

191 °C/50 mmHg (lit.)

mp

100-103 °C (lit.)
100-103 °C

SMILES string

Oc1cccc(C=O)c1

InChI

1S/C7H6O2/c8-5-6-2-1-3-7(9)4-6/h1-5,9H

InChI key

IAVREABSGIHHMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Suthan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 87, 194-198 (2011-12-24)
Novel organic nonlinear optical (NLO) material 3-hydroxybenzaldehyde (3HBA) single crystals have been grown by modified vertical Bridgman technique using the single and double wall ampoules with nano translation. Single crystal X-ray diffraction analysis reveals that 3HBA crystallizes in orthorhombic system
Yan-Chao Wu et al.
Journal of the American Chemical Society, 130(22), 7148-7152 (2008-05-06)
(-)-Quinocarcin (1) has been synthesized in a longest linear sequence of 22 steps from 3-hydroxybenzaldehyde in 16% overall yield. The Pictet-Spengler reaction of L-tert-butyl-2-bromo-5-hydroxy phenylalanate (17), synthesized according to Corey-Lygo's enantioselective alkylation process, with benzoxyacetaldehyde (12) under mild acidic conditions
B A Stamboliyska et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(7), 1541-1548 (2003-04-26)
The spectral and structural changes, caused by the conversion of the 3-hydroxybenzaldehyde molecule into the corresponding oxyanion have been studied by IR spectra, and MP2 and DFT force field calculations. The conversion causes a 13 cm(-1) decrease in the frequency
M Parekh et al.
Letters in applied microbiology, 22(2), 115-120 (1996-02-01)
Desulfovibrio desulfuricans ATCC 27774 was screened for reactivity against aromatic compounds during lactate-dependent, nitrate-dissimilating growth. Only aromatic aldehydes (benzaldehyde, 2-hydroxybenzaldehyde, 3-hydroxybenzaldehyde, 4-hydroxybenzaldehyde, vanillin, iso-vanillin and o-vanillin) were reactive and, with the exception of 2-hydroxybenzaldehyde, were stimulatory to lactate-dependent growth. Aromatic
Jayashree Yenagi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(5), 1025-1033 (2010-10-05)
Ab initio conformers and dimers have been computed at RHF and B3LYP/6-31G* levels for isomers 2-chloro-3-hydroxybenzaldehyde and 3-chloro-4-hydroxybenzaldehyde to explain the observed infrared absorption and Raman vibrational spectral features in the region 3500-50 cm(-1). The position of the chlorine in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service