Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

17152

Millipore

Karmali Campylobacter Agar (Base)

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
charcoal, 4 g/L
corn starch, 1 g/L
peptone, special, 23 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

environmental
food and beverages
veterinary

microbiology

suitability

selective for Campylobacter spp.

Application

Blood free medium for selective isolation and cultivation of thermotolerant Campylobacter species from food and animal feeds. It is recommended by ISO Committee under the specification ISO 10272:1995.

Preparation Note

Suspend 22.5 g in 490 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 50°C. Aseptically add 5 ml Hemin solution (1.6mg (Cat. No. 51280) /5ml) and rehydrated contents of 1 vial of Campylobacter Selective Supplement (Cat. No. 17780). Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shafiq M Syed et al.
Cell stem cell, 26(1), 64-80 (2019-12-31)
The remarkable regenerative capacity of the endometrium (the inner lining of the uterus) is essential for the sustenance of mammalian life. Over the years, the role of stem cells in endometrial functions and their pathologies has been suggested; however, the

Articles

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service