01769
Folic acid
Synonym(s):
Acidum folicum, PteGlu, Pteroyl-L-glutamic acid, Vitamin M
About This Item
Recommended Products
biological source
synthetic
agency
USP/NF
assay
96.0-102.0% anhydrous basis
form
powder
color
yellow
mp
250 °C
solubility
cold water: very slightly soluble 0.0016 mg/ml at 25 degrees C
boiling water: soluble 1%
1-butanol: partially soluble
acetone: insoluble
benzene: insoluble
chloroform: insoluble
diethyl ether: insoluble
dilute HCl: soluble (Hot)
ethanol: partially soluble
methanol: slightly soluble
sulfuric acid: soluble
storage temp.
2-8°C
SMILES string
NC(N1)=NC(C2=C1N=CC(CNC3=CC=C(C(N[C@@H](CCC(O)=O)C(O)=O)=O)C=C3)=N2)=O
InChI
1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1
InChI key
OVBPIULPVIDEAO-LBPRGKRZSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Preparation Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service