Z163422
Dri-Can® desiccating canister
L × W × H 4 in. × 2.06 in. × 0.53 in.
About This Item
Recommended Products
manufacturer/tradename
Multisorb
L × W × H
4 in. × 2.06 in. × 0.53 in.
General description
High Moisture Capacity
The desiccant material within Dri-Can attracts water vapor and holds it physically through surface adsorption and capillary condensation. It has a tremendous number of microscopic pores which give it a high moisture capacity. One Dri-Can will maintain a safe level of humidity in three cubic feet of sealed space.
Easy to Monitor and Regenerate
Available in a metal or plastic case, Dri-Can has either a monitor window in the metal case or a see-through, translucent plastic case. When the desiccant turns pink, it indicates that a saturation point has been reached. Regeneration can be achieved by microwave or conventional oven.
- Conventional Oven - (Metal or plastic case) - Heat Dri-Can in vented oven at 149°C (300°F) for three hours.
- Microwave Oven - stand DriCan on its side - Heat Dri-Can in microwave on high (using a 900 watt microwave) in 3 minute intervals until beads turn blue. Time may vary depending on wattage of the microwave used to regenerate.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)