W368709
2-Methoxyphenyl acetate
≥98%, FG
Synonym(s):
O-Acetylguaiacol
About This Item
biological source
synthetic
grade
FG
reg. compliance
EU Regulation 1334/2008 & 872/2012
assay
≥98%
refractive index
n20/D 1.514 (lit.)
bp
122-124 °C/18 mmHg (lit.)
density
1.129 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
documentation
see Safety & Documentation for available documents
food allergen
no known allergens
organoleptic
musty; woody; walnut
SMILES string
COc1ccccc1OC(C)=O
InChI
1S/C9H10O3/c1-7(10)12-9-6-4-3-5-8(9)11-2/h3-6H,1-2H3
InChI key
BHJHPYFAYGAPLS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service