Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N16207

Sigma-Aldrich

trans-2-Nitrocinnamaldehyde

97%

Synonym(s):

trans-3-(2-Nitrophenyl)-2-propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH=CHCHO
CAS Number:
Molecular Weight:
177.16
Beilstein/REAXYS Number:
1073058
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

127-129 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\c1ccccc1[N+]([O-])=O

InChI

1S/C9H7NO3/c11-7-3-5-8-4-1-2-6-9(8)10(12)13/h1-7H/b5-3+

InChI key

VMSMELHEXDVEDE-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Masoudiasl et al.
Materials science & engineering. C, Materials for biological applications, 61, 809-823 (2016-02-04)
Some new five coordinated ZnLX2 complexes, where L is N3-Schiff base ligand obtained by condensation reaction between diethylenetriamine and (E)-3-(2-nitrophenyl)acrylaldehyde and X (Cl(-), Br(-), I(-), N3(-) and NCS(-)), were synthesized and characterized by FT-IR, (1)H and (13)CNMR, UV-visible, ESI-mass spectra

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service