Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C119008

Sigma-Aldrich

Cyclopropyl 4-fluorophenyl ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COC3H5
CAS Number:
Molecular Weight:
164.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

119-120 °C/13 mmHg (lit.)

density

1.144 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(cc1)C(=O)C2CC2

InChI

1S/C10H9FO/c11-9-5-3-8(4-6-9)10(12)7-1-2-7/h3-7H,1-2H2

InChI key

MHKHJIJXMVHRAJ-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

212.0 °F - closed cup

flash_point_c

100.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service