Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C100757

Sigma-Aldrich

trans-1,4-Cyclohexanedicarboxylic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CO2H)2
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

>300 °C (lit.)

SMILES string

OC(=O)[C@@H]1CC[C@H](CC1)C(O)=O

InChI

1S/C8H12O4/c9-7(10)5-1-2-6(4-3-5)8(11)12/h5-6H,1-4H2,(H,9,10)(H,11,12)/t5-,6-

InChI key

PXGZQGDTEZPERC-IZLXSQMJSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luca Fusaro et al.
Biomolecules, 10(12) (2020-12-04)
In the field of artificial prostheses for damaged vessel replacement, polymeric scaffolds showing the right combination of mechanical performance, biocompatibility, and biodegradability are still demanded. In the present work, poly(butylene-co-triethylene trans-1,4-cyclohexanedicarboxylate), a biodegradable random aliphatic copolyester, has been synthesized and
Franco Dominici et al.
Carbohydrate polymers, 246, 116631-116631 (2020-08-05)
Fully biobased blends of thermoplastic starch and a poly(butylene cyclohexanedicarboxylate)-based random copolyester containing 25 % of adipic acid co-units (PBCEA) are prepared by melt blending and direct extrusion film casting. The obtained films are characterized from the physicochemical and mechanical
Adam Al-Janabi et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(8), 697-701 (2020-02-23)
The nematic twist-bend (NTB ) liquid crystal phase possesses a local helical structure with a pitch length of a few nanometres and is the first example of spontaneous symmetry breaking in a fluid system. All known examples of the NTB
Pavel A Demakov et al.
Materials (Basel, Switzerland), 13(2) (2020-01-23)
Five trans-1,4-cyclohexanedicarboxylate (chdc2-) metal-organic frameworks of transition metals were synthesized in aqueous systems. A careful control of pH, reaction temperature and solvent composition were shown to direct the crystallization of a particular compound. Isostructural [Co(H2O)4(chdc)]n (1) and [Fe(H2O)4(chdc)]n (2) consist

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service