Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900159

Sigma-Aldrich

UV-Vis light curable resin

adhesible to plastics and glass, low viscosity

Synonym(s):

CPS 1030 UVvis-A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

viscosity

82 cP (Brookfield digital viscometer with size 31 spindle)

density

1.16 g/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Storage:
  • The storage temperature should not exceed 35°C.
  • Keep tightly closed. Store in a cool, dry, well-ventilated area.
  • Do not expose to ultraviolet light.
  • High temperatures (>80°C), pressure and contamination with peroxides may result in auto-polymerization of the product.
  • Unstable upon depletion of inhibitor.

Application

  • UV curable very low viscosity resin.
  • Cures rapidly to form a glassy, high modulus material.
  • Excellent for use in applications requiring low viscosity, high hardness, and low shrinkage stress.
  • Good for some spray coatings requiring no volatile organic compounds (VOCS).
  • Non-yellowing.
  • Low oxygen inhibition.
  • Mechanical properties after curing:
Modulus*: 470 MPa
Strength*: 17.3MPa
Elongation*: 11.1%
Tack Free**: 10 fpm

*Test Resources: tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width
** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material Data using ASTM D 638-02a.

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service