806919
Titanium doped silica
mesoporous SBA-15, <150 μm particle size, pore size 4 nm, Hexagonal pore morphology
Synonym(s):
Porous Silica, SBA-15, Silicon dioxide
About This Item
Recommended Products
form
powder
concentration
3-5% (Ti content)
surface area
500-700 m2/g
avg. part. size
≤150 μm
particle size
<150 μm
pore size
0.5-0.7 cm3/g pore volume
3-5 μm pore diameter
4 nm pore size
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Mesoporous silica used in wastewater remediation, air cleaning, catalysis, bio-catalysis, and drug delivery.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service