Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769541

Sigma-Aldrich

Strontium hydroxide octahydrate

≥97%

Synonym(s):

Strontium dihydroxide octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sr(OH)2 · 8H2O
CAS Number:
Molecular Weight:
265.76
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.23
assay:
≥97%
97.0-103.0% (T)
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

assay

≥97%
97.0-103.0% (T)

form

solid

impurities

≤0.02% total nitrogen
≤2.0% carbonate (as SrCO3)

density

1.9 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%

cation traces

Ba: ≤0.1%
Ca: ≤0.05%
Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

SMILES string

O.O.O.O.O.O.O.O.O[Sr]O

InChI

1S/10H2O.Sr/h10*1H2;/q;;;;;;;;;;+2/p-2

InChI key

UJPWWRPNIRRCPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Strontium hydroxide octahydrate is used as precursor material for synthesis of ceramics and oxides having applications in solid oxide fuel cells (SOFC) and photocatalysis.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical charge transfer observed in nanoscale hydrogen evolving photocatalysts using surface photovoltage spectroscopy
Wang, J., et al.
Energy & Environmental Science, 8, 2970-2976 (2015)
Preparation of nanoscale composite LSCF/GDCS cathode materials by microwave sintering for intermediate-temperature SOFC applications
Hua, CH. and Chou, CC.
Ceramics International, 41, S708-S712 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service