Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

748366

Sigma-Aldrich

(R)-1-(Diphenylphosphino)-3-methyl-2-butylamine

97%

Synonym(s):

(R)-1-(Diphenylphosphino)-3-methylbutan-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22NP
CAS Number:
Molecular Weight:
271.34
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.597

density

1.049 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)[C@@H](N)CP(c1ccccc1)c2ccccc2

InChI

1S/C17H22NP/c1-14(2)17(18)13-19(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,14,17H,13,18H2,1-2H3/t17-/m0/s1

InChI key

ZZLCXURCZWQECA-KRWDZBQOSA-N

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service