Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698547

Sigma-Aldrich

α-(N,N-Dimethylamino)ethylferrocene

97%

Synonym(s):

N,N-Dimethyl-1-ferrocenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H5)Fe(C5H4CH(CH3)N(CH3)2)
CAS Number:
Molecular Weight:
257.15
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

reaction suitability

reagent type: catalyst
core: iron

refractive index

n20/D 1.587

density

1.239 g/mL at 25 °C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CC([C]2[CH][CH][CH][CH]2)N(C)C

InChI

1S/C9H14N.C5H5.Fe/c1-8(10(2)3)9-6-4-5-7-9;1-2-4-5-3-1;/h4-8H,1-3H3;1-5H;

InChI key

ISOLNZQTVMCEOI-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Frustrated Lewis pairs
  • Substituted ferrocenecarboxylic acids and esters

Reactant for polymer-supported ferrocene derivatives

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service