Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

690473

Sigma-Aldrich

1-(2-Hydroxyethyl)-3-methylimidazolium dicyanamide

97%, ≥96.5% (HPLC)

Synonym(s):

[C2OHMIM][DCA]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N5O
CAS Number:
Molecular Weight:
193.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥96.5% (HPLC)
97%

form

liquid

impurities

≤1.0% water

functional group

amine
hydroxyl

SMILES string

N#C[N-]C#N.C[n+]1ccn(CCO)c1

InChI

1S/C6H11N2O.C2N3/c1-7-2-3-8(6-7)4-5-9;3-1-5-2-4/h2-3,6,9H,4-5H2,1H3;/q+1;-1

InChI key

UBQLCZUQCFNAGM-UHFFFAOYSA-N

Application

1-(2-Hydroxyethyl)-3-methylimidazolium dicyanamide or [EtOHmim][DCA] is an ionic liquid that can be used:
  • As a solvent in the study of absorption of ammonia.[1]
  • As a polymerization initiator, which is known to accelerate the cure of cyanate ester resins.[2]
  • In the thermodynamic study of the solubility of D-sorbitol, xylitol, and meso-erythritol in ionic liquids.[3]
  • As a corrosion inhibitor of API 5L X52 steel in acidic conditions.[4]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance of (HEMIM)(CN)-and (DMIM)(Cl)-as corrosion inhibitor of API 5L X52 steel in HCl medium
Corrales-Luna M, et al.
International Journal of Electrochemical Science, 14, 4420-4442 (2019)
Acceleration of cyanate ester trimerization by dicyanamide RTILs
Throckmorton J and Palmese G
Polymer, 91, 7-13 (2016)
Screening ionic liquids as suitable ammonia absorbents on the basis of thermodynamic and kinetic analysis
Bedia J, et al.
Separation and Purification Technology, 95, 188-195 (2012)
Solid-liquid phase equilibria in binary mixtures of functionalized ionic liquids with sugar alcohols: New experimental data and modelling
Paduszynski K, et al.
Fluid Phase Equilibria, 403, 167-175 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service