Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

688681

Sigma-Aldrich

Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphonium tetrafluoroborate

95%

Synonym(s):

Dicyclohexyl[9-(3-phenylpropyl)-9-fluorenyl]phosphine tetrafluoroborate, cataCXium® FPrPh

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H42BF4P
CAS Number:
Molecular Weight:
568.48
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

assay

95%

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

F[B-](F)(F)F.C1CCC(CC1)[PH+](C2CCCCC2)C4(CCCc3ccccc3)c5ccccc5-c6ccccc46

InChI

1S/C34H41P.BF4/c1-4-15-27(16-5-1)17-14-26-34(32-24-12-10-22-30(32)31-23-11-13-25-33(31)34)35(28-18-6-2-7-19-28)29-20-8-3-9-21-29;2-1(3,4)5/h1,4-5,10-13,15-16,22-25,28-29H,2-3,6-9,14,17-21,26H2;/q;-1/p+1

InChI key

VBCMIQSEFSKZNU-UHFFFAOYSA-O

General description

sold in collaboration with Solvias AG

Legal Information

cataCXium is a registered trademark of Umicore AG & Co. KG

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph A Fleckenstein et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(14), 4267-4279 (2008-03-28)
A dicyclohexyl(2-sulfo-9-(3-(4-sulfophenyl)propyl)-9H-fluoren-9-yl)phosphonium salt was synthesized in 64% overall yield in three steps from simple commercially available starting materials. The highly water-soluble catalyst obtained from the corresponding phosphine and [Na(2)PdCl(4)] enabled the Suzuki coupling of a broad variety of N- and
C. A. Fleckenstein, et al.,
Organic Process Research & Development, 12, 475-479 (2008)
Aqueous cross-coupling: highly efficient Suzuki-Miyaura coupling of N-heteroaryl halides and N-heteroarylboronic acids.
Fleckenstein, C. A.; Plenio, H.
Green Chemistry, 9, 1287-1287 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service