Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

683949

Sigma-Aldrich

7-Hydroxy-4-chromone

97%

Synonym(s):

7-Hydroxy-4H-1-benzopyran-4-one, 7-Hydroxybenzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

215-220 °C

functional group

ketone

SMILES string

Oc1ccc2C(=O)C=COc2c1

InChI

1S/C9H6O3/c10-6-1-2-7-8(11)3-4-12-9(7)5-6/h1-5,10H

InChI key

WVJCRTSTRGRJJT-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Truong et al.
SLAS technology, 26(3), 287-299 (2020-12-10)
Derivation and differentiation of human induced pluripotent stem cells (hiPSCs) provide the opportunity to generate medically important cell types from individual patients and patient populations for research and the development of potential cell therapies. This technology allows disease modeling and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service