Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

683868

Sigma-Aldrich

3-Aminocarbonylphenylboronic acid

Synonym(s):

3-Carbamoylphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2NCO)C6H4B(OH)2
CAS Number:
Molecular Weight:
164.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

mp

221-226 °C

functional group

amide

storage temp.

2-8°C

SMILES string

NC(=O)c1cccc(c1)B(O)O

InChI

1S/C7H8BNO3/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4,11-12H,(H2,9,10)

InChI key

WDGWHKRJEBENCE-UHFFFAOYSA-N

Application

Reactant involved in the synthesis of a variety of inhibitors including:
  • Orally active phosphodiesterase 10A inhibitors
  • Pyrimidine derivatives as TpI2 kinase inhibitors
  • Peptidomimetic inhibitors of STAT3 protein

Reactant involved in:
  • Oxidative cross-coupling with mercaptoacetylenes
  • Trifluoromethylation via a Collidine trifluoromethylating agent
  • Suzuki cross-coupling reactions

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service