Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

682780

Sigma-Aldrich

Poly(3-dodecylthiophene-2,5-diyl)

electronic grade, 99.995% trace metals basis, average Mw ~27,000

Synonym(s):

P3DDT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

grade

electronic grade

description

Band gap: 1.74 eV
regioregular

assay

99.995% trace metals basis

form

solid

mol wt

average Mw ~27,000

transition temp

Tm (DSC) 165.6 °C (peak)

orbital energy

HOMO -5.29 eV 
LUMO -3.55 eV 

OPV device performance

ITO/PEDOT:PSS/P3DDT/PC61BM (1:3)/LiF/Al

  • Short-circuit current density (Jsc): 2.9 mA/cm2
  • Open-circuit voltage (Voc): 0.6 V
  • Fill Factor (FF): 0.38
  • Power Conversion Efficiency (PCE): 0.65 %

semiconductor properties

P-type (mobility=10−4 - 10−1 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-dodecylthiophene-2,5-diyl) (P3DDT) is a poly(3-alkylthiophene) based conducting polymer that shows reversible electrochemistry with high conductivity. It is a regioregular polymer that can be used in the development of a variety of organic electronics based devices.

Application

Conducting polymer.
Rechargeable battery electrodes, electrochromic devices, chemical and optical sensors, light-emitting diodes, microelectrical amplifiers, field-effect transistors and non-linear optical materials.
P3DDT can be used as a conjugating polymer in the fabrication of electrochemical devices which include polymeric solar cells, chemical sensors and field effect transistors.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagamatsu S., et al.
Applied Physics Letters, 84, 4608-4608 (2004)
Inkjet printed chemical sensor array based on polythiophene conductive polymers
Li B, et al.
Sensors and Actuators B, Chemical, 123(2), 651-660 (2007)
Evaluation of solution-processable carbon-based electrodes for all-carbon solar cells
Ramuz MP, et al.
ACS Nano, 6(11), 10384-10395 (2012)
Self-orienting head-to-tail poly (3-alkylthiophenes): new insights on structure-property relationships in conducting polymers
McCullough RD, et al.
Journal of the American Chemical Society, 115(11), 4910-4911 (1993)
Carbon nanotube network ambipolar field-effect transistors with 108 on/off ratio
Derenskyi V, et al.
Advanced Materials, 26(34), 5969-5975 (2014)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professor Chen (Nankai University, China) and his team explain the strategies behind their recent record-breaking organic solar cells, reaching a power conversion efficiency of 17.3%.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service