672645
3-Phenyl-2-propynenitrile
98%
Synonym(s):
Phenylpropiolonitrile
About This Item
Recommended Products
assay
≥97.5% (GC)
98%
mp
37-42 °C
storage temp.
2-8°C
SMILES string
N#CC#Cc1ccccc1
InChI
1S/C9H5N/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6H
InChI key
IYXVSRXFGYDNEV-UHFFFAOYSA-N
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service