Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

667978

Sigma-Aldrich

Gold Surface Cleaning solution

Synonym(s):

Gold surface prep solution, Silver Surface Cleaning Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

Application

Recommended for cleaning metal films immediately prior to further processing. Gold/Silver Cleaner is an effective surface activator and adhesion promoter for gold and silver substrates prior to bonding processes.

Features and Benefits

Enhances fluxing, soldering, etching, plating, and coating, No adverse electrical effects, Cyanide-free. Use in Pyrex glass, fiberglass, or polypropylene tank between 20-50 °C. Immerse surface for 30-60 seconds or as needed and rinse with DI water

pictograms

Corrosion

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service