Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

666424

Sigma-Aldrich

Methyl 3-(4-methoxyphenyl)propionate

97%

Synonym(s):

Methyl p-methoxyhydrocinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H4CH2CH2CO2CH3
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

bp

120 °C

mp

37-41 °C

SMILES string

COC(=O)CCc1ccc(OC)cc1

InChI

1S/C11H14O3/c1-13-10-6-3-9(4-7-10)5-8-11(12)14-2/h3-4,6-7H,5,8H2,1-2H3

InChI key

AKQLYAFBUYHFCK-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service