Skip to Content
MilliporeSigma
All Photos(2)

Documents

645591

Sigma-Aldrich

4-Fluorocinnamoyl chloride

97%, predominantly trans

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH=CHCOCl
CAS Number:
Molecular Weight:
184.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

bp

90 °C/0.1 mmHg (lit.)

mp

42-46 °C (lit.)

SMILES string

[H]\C(=C(\[H])c1ccc(F)cc1)C(Cl)=O

InChI

1S/C9H6ClFO/c10-9(12)6-3-7-1-4-8(11)5-2-7/h1-6H/b6-3+

InChI key

GLBIKPKWQDIQCJ-ZZXKWVIFSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

>230.0 °F - closed cup

flash_point_c

> 110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service