Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

645567

Sigma-Aldrich

11-Bromoundecanamide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$40.20

$40.20

List Price$80.40Save 50%

Available to ship onMarch 28, 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
$40.20

About This Item

Linear Formula:
Br(CH2)10CONH2
CAS Number:
Molecular Weight:
264.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$40.20

List Price$80.40Save 50%

Available to ship onMarch 28, 2025Details


Request a Bulk Order

assay

97%

form

solid

mp

88-92 °C (lit.)

functional group

amide
bromo

SMILES string

NC(=O)CCCCCCCCCCBr

InChI

1S/C11H22BrNO/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10H2,(H2,13,14)

InChI key

NHUWVMSJSKNFHL-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service