Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

594415

Sigma-Aldrich

Poly[(m-phenylenevinylene)-alt-(2-methoxy-5-octyloxy-p-phenylenevinylene)]

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

solubility

chloroform: soluble
methanol: insoluble

λmax

399 nm

fluorescence

λex 402 nm; λem 450 nm in chloroform

General description

Semi-conjugated light emitting polymer.

Application

Used in the purification of carbon nanotubes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service