Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

587907

Sigma-Aldrich

3,5-Dimethylphenol-2,4,6-d3

98 atom % D

Synonym(s):

2,4,6-Trideuterio-3,5-dimethylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6D3OH
CAS Number:
Molecular Weight:
125.18
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

solid

bp

222 °C (lit.)

mp

65-66 °C (lit.)

mass shift

M+3

SMILES string

[2H]c1c(C)c([2H])c(O)c([2H])c1C

InChI

1S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3/i3D,4D,5D

InChI key

TUAMRELNJMMDMT-QGZYMEECSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service