482846
(S,S)-(−)-1,2-Di(1-naphthyl)-1,2-ethanediol
98%
About This Item
Recommended Products
assay
98%
optical activity
[α]23/D −25°, c = 1 in chloroform
mp
149-151 °C (lit.)
SMILES string
O[C@H]([C@@H](O)c1cccc2ccccc12)c3cccc4ccccc34
InChI
1S/C22H18O2/c23-21(19-13-5-9-15-7-1-3-11-17(15)19)22(24)20-14-6-10-16-8-2-4-12-18(16)20/h1-14,21-24H/t21-,22-/m0/s1
InChI key
MNRCSHSTIWDUPJ-VXKWHMMOSA-N
Application
- A ligand in the asymmetric sulfoxidation of dihydrothienopyrimidine.[1]
- A reactant for the preparation of (2S,2′S)-[(1S,2S)-1,2-bis(naphthalen-1-yl)ethane-1,2-diyl] bis[2-(dimethylamino)propanoate], a chiral bisester ligand via condensation reaction with N,N-dimethylamino acid using aldol-Tishchenko condensation catalyst.[2]
- A reactant to synthesize diol-SnCl4 chiral Bronsted acid complex, which is employed as a catalyst in the allylboration of aldehydes.[3]
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service