Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

464198

Sigma-Aldrich

Zirconyl chloride solution

30% in hydrochloric acid

Synonym(s):

Dichlorooxozirconium, Oxozirconium(IV) chloride, Zirconium dichloride monoxide, Zirconium oxide dichloride, Zirconium oxychloride, Zirconyl chloride, Zirconyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrOCl2
CAS Number:
Molecular Weight:
178.13
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
liquid

form

liquid

concentration

10-15% (titration by NaOH, % HCl)
12.8-17.9% Zr (gravimetric)
30% in hydrochloric acid

mp

−15 °C

density

1.344 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

Cl[Zr](Cl)=O

InChI

1S/2ClH.O.Zr/h2*1H;;/q;;;+2/p-2

InChI key

IPCAPQRVQMIMAN-UHFFFAOYSA-L

Application

Building block for a new series of mixed zirconium phosphonates which may have uses in catalysis, ion exchange or photophysics.[1]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reza Hosseinzadeh et al.
Scientific reports, 9(1), 14899-14899 (2019-10-19)
Pharmaceutical applications of methylene blue, especially as photosensitizer, have been limited due to its rapid enzymatic reduction in the biological systems. In this study nano-platelet zirconium phosphate was synthesized and its biocompatibility was evaluated. The synthesized material was considered as
Manuel Checa et al.
Nanomaterials (Basel, Switzerland), 9(4) (2019-04-17)
The influence of boron, tungsten and molybdenum modifiers on zirconia-based Pt catalyst was studied for glycerol valorization. Zirconia modified supports were prepared by impregnation of ZrO₂ with either boric, silicontungstic or phosphomolybdic acids to obtain supports with enhanced Brönsted acidic
Leenstra, W.R. Amicangelo, J.C.
Inorganic Chemistry, 37, 5317-5317 (1998)
Y Nayak et al.
Journal of materials science. Materials in medicine, 19(6), 2437-2444 (2008-01-26)
Hydroxyapatite (HA)-TZP (2.5 mol% Y2O3) containing 2, 5, 7.5 and 10 wt% TZP were prepared using calcium nitrate, diammonium hydrogen orthophosphate, zirconium oxychloride and yttrium nitrate. The composite powder was prepared by a reverse strike precipitation method at a pH
D R Mulinari et al.
Carbohydrate research, 345(13), 1865-1871 (2010-07-06)
Surface modification of natural fibers has been made using different methods. In this paper, cellulose fibers from sugarcane bagasse were bleached and modified by zirconium oxychloride in situ. The chemically modified cellulose fibers were compared to those of bleached ones.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service