Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

259306

Sigma-Aldrich

3-Bromothiophenol

95%

Synonym(s):

3-Bromobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4SH
CAS Number:
Molecular Weight:
189.07
Beilstein/REAXYS Number:
1926242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.634 (lit.)

bp

123-124 °C/40 mmHg (lit.)

density

1.832 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Sc1cccc(Br)c1

InChI

1S/C6H5BrS/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

HNGQQUDFJDROPY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Bromothiophenol has been used in the preparation of:
  • 3-lithium-O-lithiophenoxide[1]
  • bis[(diarylamino)styryl]benzene functionalized with sulfonium groups[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and application of high-sensitivity two-photon initiators for three-dimensional microfabrication.
Kuebler SM, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 158(2), 163-170 (2003)
Preparation and trapping of 3-lithium-O-lithiophenoxide.
Selnick HG, et al.
Tetrahedron Letters, 34(13), 2043-2046 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service