Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21935

Sigma-Aldrich

(5-Carboxypentyl)triphenylphosphonium bromide

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P(Br)(CH2)5COOH
CAS Number:
Molecular Weight:
457.34
Beilstein/REAXYS Number:
4649483
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

≥98.0%

mp

197-201 °C

SMILES string

[Br-].OC(=O)CCCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of inhibitor of protein tyrosine phosphatase 1B for treatment of diabetes and obesity
  • Synthesis of folate receptor-specific glycinamide ribonucleotide formyltransferase inhibitors with antitumor activity
  • Preparation of peptide nucleic acids (PNA) with high specific activity
  • Synthesis of roseophilin via Wittig/aldol methodology

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service