Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

197297

Sigma-Aldrich

Ethyl 1-bromocyclobutanecarboxylate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC4H6CO2C2H5
CAS Number:
Molecular Weight:
207.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

form

liquid

refractive index

n20/D 1.471 (lit.)

bp

85-88 °C/12 mmHg (lit.)

density

1.279 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C1(Br)CCC1

InChI

1S/C7H11BrO2/c1-2-10-6(9)7(8)4-3-5-7/h2-5H2,1H3

InChI key

UTVNSHXHFRIXMM-UHFFFAOYSA-N

Application

Ethyl 1-bromocyclobutanecarboxylate was used in the synthesis of 1−aminocyclobutane carboxylic acid[1].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Procedure for Preparation of C−and N−Protected 1−Aminocyclobutane Carboxylic Acid.
Synthetic Communications, 34(4), 607-613 (2004)
Daria Kupczyk et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-19)
Glucocorticoid metabolism at the tissue level is regulated by two isoenzymes 11β-hydroxysteroid dehydrogenase (11β-HSD), which mutually convert biologically active cortisol and inactive cortisone. Recent research is focused on the role of 11β-HSD1 and 11β-HSD2 as autocrine factors of tumor cell
Renata Studzińska et al.
Chemistry & biodiversity, 16(6), e1900065-e1900065 (2019-04-24)
11β-Hydroxysteroid dehydrogenase type 1 (11β-HSD1) is an enzyme that affects the body's cortisol levels. The inhibition of its activity can be used in the treatment of Cushing's syndrome, metabolic syndrome and type 2 diabetes. In this study, we synthesized new

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service